Anti-MYC Rabbit Polyclonal Antibody (Biotin)

BS-4963R-BIO
10405-188EA 545.21 USD
10405-188
Anti-MYC Rabbit Polyclonal Antibody (Biotin)
Transcription factor that binds DNA in a non-specific manner, yet also specifically recognizes the core sequence 5'-CAC[GA]TG-3'. Activates the transcription of growth-related genes.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: MYC
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR