Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF10D Rabbit Polyclonal Antibody (Biotin)

Supplier: Bioss
BS-4962R-BIO
Receptor for the cytotoxic ligand TRAIL. Contains a truncated death domain and hence is not capable of inducing apoptosis but protects against TRAIL-mediated apoptosis. Reports are contradictory with regards to its ability to induce the NF-kappa-B pathway. According to PubMed:9382840, it cannot but according to PubMed:9430226, it can induce the NF-kappa-B pathway.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: TNFRSF10D
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-4962R-BIO 10405-164 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TNFRSF10D
Antigen Name Tumor Necrosis Factor Receptor Superfamily Member 10D
Reactivity Human
Conjugation Biotin
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms TRUNDD, TRAIL-R4, CD264, DCR2, TRAILR4
Gene ID 8793
Concentration 1 μg/μl
Cross Adsorption No
Immunogen 160-210/386
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty