Anti-CD1D1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-4961R-CY5
10405-160EA 811.1 USD
10405-160
Anti-CD1D1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Antigen-presenting protein that binds self and non-self glycolipids and presents them to T-cell receptors on natural killer T-cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD1D1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR