Anti-CD1D1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-4961R-A647
10399-722EA 811.1 USD
10399-722
Anti-CD1D1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Antigen-presenting protein that binds self and non-self glycolipids and presents them to T-cell receptors on natural killer T-cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD1D1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR