Anti-PCGF1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-4958R-A555
10399-710EA 811.1 USD
10399-710
Anti-PCGF1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
PCGF1 is a component of the Polycomb group (PcG) multiprotein BCOR complex, a complex required to maintain the transcriptionally repressive state of some genes, such as BCL6 and the cyclin-dependent kinase inhibitor, CDKN1A. It represses CDKN1A expression by binding to its promoter, and this repression is dependent on the retinoic acid response element (RARE element). PCGF1 als promotes cell cycle progression and also enhances cell proliferation, thus it may have a positive role in tumor cell growth.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCGF1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR