Anti-CRTAM Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-4957R-CY5
10399-694EA 811.1 USD
10399-694
Anti-CRTAM Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Interaction with CADM1 promotes natural killer (NK) cell cytotoxicity and interferon-gamma (IFN-gamma) secretion by CD8+ cells in vitro as well as NK cell-mediated rejection of tumors expressing CADM3 in vivo.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRTAM
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR