|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | CRTAM |
Antigen Name | Cytotoxic And Regulatory T Cell Molecule |
Reactivity | Mouse, Human, Rat |
Conjugation | Biotin |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ImmunoChemistry | Yes |
Antigen Synonyms | CD355, Class-I MHC-restricted T-cell-associated molecule, Cytotoxic and regulatory T-cell molecule, CRTAM |
Gene ID | 56253 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Immunogen | 40-90/393 |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |