Anti-CRTAM Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-4957R-A350
10399-682EA 811.1 USD
10399-682
Anti-CRTAM Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Interaction with CADM1 promotes natural killer (NK) cell cytotoxicity and interferon-gamma (IFN-gamma) secretion by CD8+ cells in vitro as well as NK cell-mediated rejection of tumors expressing CADM3 in vivo.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRTAM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR