Anti-ERAS Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-4950R-CY3
10405-148EA 811.1 USD
10405-148
Anti-ERAS Rabbit Polyclonal Antibody (Cy3®)
Antibodies
ERAS belongs to the small GTPase superfamily. Ras family. Ras proteins bind GDP/GTP and possess intrinsic GTPase activity. The ERas gene is expressed in embryonic stem (ES) cells and promotes their in vitro proliferation and tumorigenicity. ERas-null ES cells maintain pluripotency but show significantly reduced growth and tumorigenicity, which are rescued by expression of ERas complementary DNA or by activated phosphatidylinositol-3-OH kinase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ERAS
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR