Anti-STFA3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-4937R-A555
10405-056EA 811.1 USD
10405-056
Anti-STFA3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Cystatin A and cystatin B (also designated PME, CST6, STFB, CPI-B, stefin B and liver thiol proteinase inhibitor) are thiol protease inhibitors that form complexes with papain and the cathepsins B, H, and L. Cystatin A, a cytoplasmic protein, is one of the precursor proteins of the cornified cell envelope in keratinocytes and plays a role in epidermal development and maintenance. Cystatin B protects against intracellular proteases leaking out of lysosomes and is primarily expressed in heart, liver and kidney.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STFA3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR