Anti-Escherichka coli K99 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-4920R-HRP
10405-048EA 811.1 USD
10405-048
Anti-Escherichka coli K99 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
K88-K99 minor fimbrial subunit, plays an essential role in the biogenesis of the K99 fimbriae. required at some step in the initiation and/or elongation of the K99 fimbriae.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: Escherichka coli K99
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR