|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | CD44 |
Antigen Name | CD44 molecule (Indian blood group) |
Reactivity | Mouse, Human, Rat |
Conjugation | Alexa Fluor® 488 |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | MC56, LHR, ECMR-III, MDU2, HCELL, Pgp1, MIC4, CDW44, IN, MDU3, CSPG8, HUTCH-I |
Gene ID | 960 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Immunogen | 230-280/742 |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |