Anti-CSF1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-4910R-A647
10404-980EA 811.1 USD
10404-980
Anti-CSF1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Cytokine that plays an essential role in the regulation of survival, proliferation and differentiation of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes. Promotes the release of proinflammatory chemokines, and thereby plays an important role in innate immunity and in inflammatory processes. Plays an important role in the regulation of osteoclast proliferation and differentiation, the regulation of bone resorption, and is required for normal bone development. Required for normal male and female fertility. Promotes reorganization of the actin cytoskeleton, regulates formation of membrane ruffles, cell adhesion and cell migration. Plays a role in lipoprotein clearance.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CSF1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR