Anti-NPTX1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-4893R-CY3
10405-930EA 811.1 USD
10405-930
Anti-NPTX1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
NPTX1 is a member of the neuronal pentraxin gene family. Neuronal pentraxin 1 is similar to the rat NP1 gene which encodes a binding protein for the snake venom toxin taipoxin. Human NPTX1 mRNA is exclusively localized to the nervous system.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NPTX1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR