Anti-APOE Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4892R-CY7
10400-864EA 811.1 USD
10400-864
Anti-APOE Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Mediates the binding, internalization, and catabolism of lipoprotein particles. It can serve as a ligand for the LDL (apo B/E) receptor and for the specific apo-E receptor (chylomicron remnant) of hepatic tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APOE
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR