|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | APOE |
Antigen Name | Apolipoprotein E |
Reactivity | Rat, Human, Mouse |
Conjugation | Cy3® |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | AD2, LDLCQ5, apo-E, MGC1571, LPG |
Gene ID | 348 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Immunogen | 149-180/317 |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |