Anti-CD24A Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4891R-CY7
10404-930EA 811.1 USD
10404-930
Anti-CD24A Rabbit Polyclonal Antibody (Cy7®)
Antibodies
May have a specific role to play in early thymocyte development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD24A
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR