|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | CD24 |
Antigen Name | CD24 molecule |
Reactivity | Human |
Conjugation | Alexa Fluor® 350 |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Flow Cytometry | Yes |
Antigen Synonyms | FLJ22950, CD24A, FLJ43543, MGC75043 |
Gene ID | 100133941 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Immunogen | 20-60/80 |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |