Anti-MS4A2 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-4870R-CY3
10399-194EA 811.1 USD
10399-194
Anti-MS4A2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
High affinity receptor that binds to the Fc region of immunoglobulins epsilon. Aggregation of FCER1 by multivalent antigens is required for the full mast cell response, including the release of preformed mediators (such as histamine) by degranulation and de novo production of lipid mediators and cytokines. Also mediates the secretion of important lymphokines. Binding of allergen to receptor-bound IgE leads to cell activation and the release of mediators responsible for the manifestations of allergy.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MS4A2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR