Anti-S6K1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4867R-CY7
10399-176EA 811.1 USD
10399-176
Anti-S6K1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
This gene encodes a member of the ribosomal S6 kinase family of serine/threonine kinases. The encoded protein responds to mTOR (mammalian target of rapamycin) signaling to promote protein synthesis, cell growth, and cell proliferation. Activity of this gene has been associated with human cancer. Alternatively spliced transcript variants have been observed. The use of alternative translation start sites results in isoforms with longer or shorter N-termini which may differ in their subcellular localizations. There are two pseudogenes for this gene on chromosome 17. [provided by RefSeq, Jan 2013].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: S6K1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR