Anti-RUNX3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-4848R-A488
10405-886EA 810.87 USD
10405-886
Anti-RUNX3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
RUNX3 binds to the core site of murine Leukemia virus, the core sequences in the enhancer of the polyomavirus, and also to the enhancers of the T-cell receptor genes. May be involved in the control of cellular proliferation and/or differentiation (By similarity). Heterodimer of an alpha and a beta subunit. The alpha subunit binds DNA as a monomer and through the Runt domain.DNA-binding is increased by heterodimerization. Interacts with TLE1 and SUV39H1, Subcellular location in Nucleus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RUNX3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR