Anti-F7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-4846R-A647
10399-092EA 811.1 USD
10399-092
Anti-F7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Initiates the extrinsic pathway of blood coagulation. Serine protease that circulates in the blood in a zymogen form. Factor VII is converted to factor VIIa by factor Xa, factor XIIa, factor IXa, or thrombin by minor proteolysis. In the presence of tissue factor and calcium ions, factor VIIa then converts factor X to factor Xa by limited proteolysis. Factor VIIa will also convert factor IX to factor IXa in the presence of tissue factor and calcium.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Factor VII
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR