Anti-TJP2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-4844R-FITC
10399-080EA 811.1 USD
10399-080
Anti-TJP2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Plays a role in tight junctions and adherens junctions.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TJP2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR