Anti-EIF2S1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-4842R-A555
10399-058EA 810.86 USD
10399-058
Anti-EIF2S1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Functions in the early steps of protein synthesis by forming a ternary complex with GTP and initiator tRNA. This complex binds to a 4S ribosomal subunit, followed by mRNA binding to form a 43S preinitiation complex. Junction of the 6S ribosomal subunit to form the 8S initiation complex is preceded by hydrolysis of the GTP bound to eIF-2 and release of an eIF-2-GDP binary complex. In order for eIF-2 to recycle and catalyze another round of initiation, the GDP bound to eIF-2 must exchange with GTP by way of a reaction catalyzed by eIF-2B.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EIF2S1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR