Anti-nonstructural protein 1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4839R-CY7
10404-772EA 811.1 USD
10404-772
Anti-nonstructural protein 1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza type C infections cause a mild respiratory illness and are not thought to cause epidemics. Influenza A viruses are further categorized into subtypes on the basis of two surface antigens: hemagglutinin (H) and neuraminidase (N). Strains are also described by geographic origin, strain number and year of isolation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: nonstructural protein 1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR