Anti-NT5E Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4834R-CY7
10400-734EA 811.1 USD
10400-734
Anti-NT5E Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Hydrolyzes extracellular nucleotides into membrane permeable nucleosides. Exhibits AMP-, NAD-, and NMN-nucleosidase activities.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: NT5E
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Dog, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR