Anti-CD83 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-4826R-CY5.5
10400-722EA 811.1 USD
10400-722
Anti-CD83 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
May play a significant role in antigen presentation or the cellular interactions that follow lymphocyte activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD83
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR