|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | CD4 |
Antigen Name | CD4 molecule |
Reactivity | Human |
Conjugation | Cy5® |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | Ly-4, W3/25, L3T4, p55, CD4mut |
Gene ID | 920 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Immunogen | 320-370/458 |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |