Anti-CD4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-4817R-A488
10400-678EA 811.1 USD
10400-678
Anti-CD4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Accessory protein for MHC class-II antigen/T-cell receptor interaction. May regulate T-cell activation. Induces the aggregation of lipid rafts.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD4
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR