Anti-CD3E Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-4816R-A647
10400-656EA 811.1 USD
10400-656
Anti-CD3E Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
The CD3 complex mediates signal transduction.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD3E
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR