Anti-CD3E Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4815R-CY7
10398-932EA 811.1 USD
10398-932
Anti-CD3E Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The CD3 complex mediates signal transduction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD3E
Clonality: Polyclonal
Clone: 0
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR