Anti-CD3E Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-4815R-A350
10398-916EA 810.86 USD
10398-916
Anti-CD3E Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
The CD3 complex mediates signal transduction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD3E
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR