Anti-Newcastle disease virus Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-4814R-HRP
10400-652EA 811.1 USD
10400-652
Anti-Newcastle disease virus Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Newcastle disease virus

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: Newcastle disease virus
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR