Anti-Newcastle disease virus Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-4814R-A350
10398-906EA 811.1 USD
10398-906
Anti-Newcastle disease virus Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Newcastle disease virus

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Newcastle disease virus
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR