Anti-pig cholerae salmonella Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-4804R-CY7
10400-630EA 757.82 USD
10400-630
Anti-pig cholerae salmonella Rabbit Polyclonal Antibody (Cy7®)
Antibodies
pig cholerae salmonella

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: pig cholerae salmonella
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR