Anti-pig cholerae salmonella Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-4804R-CY3
10400-624EA 746.42 USD
10400-624
Anti-pig cholerae salmonella Rabbit Polyclonal Antibody (Cy3®)
Antibodies
pig cholerae salmonella

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: pig cholerae salmonella
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR