Anti-pig cholerae salmonella Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-4804R-A488
10398-850EA 746.42 USD
10398-850
Anti-pig cholerae salmonella Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
pig cholerae salmonella

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: pig cholerae salmonella
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR