The genus Salmonella is a member of the family Enterobacteriaceae. The genus is composed of Gram-negative bacilli that are facultative and flagellated (motile). Salmonellae possess 3 major antigens; the "H" or flagellar antigen (phase 1 & 2), the "O" or somatic antigen (part of the LPS moiety) and the "Vi" or capsular antigen (referred to as "K" in other Enterobacteriaceae). Salmonellae also possess the LPS endotoxin characteristic of Gram-negative bacteria. This LPS is composed of an "O" polysaccharide ("O" antigen) an "R" core and the endotoxic inner "Lipid A". Endotoxins evoke fever and can activate complement, kinin and clotting factors. Until recently the most common cause of food poisoning by Salmonella species was due to S. Typhimurium. As its name suggests, it causes a typhoid-like disease in mice. In humans S. Typhimurium does not cause as severe disease as S. Typhi, and is not normally fatal. The disease is characterized by diarrhea, abdominal cramps, vomiting and nausea, and generally lasts up to 7 days.
Recommended Dilutions: IHC-P: 1:100-500
Type: Primary
Antigen: Salmonella typhimurium
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse