Anti-CD8A Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-4791R-A488
10398-804EA 811.1 USD
10398-804
Anti-CD8A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Identifies cytotoxic/suppressor T-cells that interact with MHC class I bearing targets. CD8 is thought to play a role in the process of T-cell mediated killing. CD8 alpha chains binds to class I MHC molecules alpha-3 domains.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD8A
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR