Anti-CD8A Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4790R-CY7
10398-800EA 811.1 USD
10398-800
Anti-CD8A Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Identifies cytotoxic/suppressor T-cells that interact with MHC class I bearing targets. CD8 is thought to play a role in the process of T-cell mediated killing. CD8 alpha chains binds to class I MHC molecules alpha-3 domains.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD8A
Clonality: Polyclonal
Clone: 0
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR