Anti-CD19 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-4787R-CY5
10398-772EA 746.43 USD
10398-772
Anti-CD19 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Assembles with the antigen receptor of B-lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD19
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR