Anti-IL6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-4587R-A350
10403-502EA 811.1 USD
10403-502
Anti-IL6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Cytokine with a wide variety of biological functions. It is a potent inducer of the acute phase response. Plays an essential role in the final differentiation of B-cells into Ig-secreting cells Involved in lymphocyte and monocyte differentiation. Acts on B-cells, T-cells, hepatocytes, hematopoietic progenitor cells and cells of the CNS. Required for the generation of T(H)17 cells. Also acts as a myokine. It is discharged into the bloodstream after muscle contraction and acts to increase the breakdown of fats and to improve insulin resistance. It induces myeloma and plasmacytoma growth and induces nerve cells differentiation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR