Anti-IL2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-4586R-A647
10403-460EA 811.1 USD
10403-460
Anti-IL2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Produced by T-cells in response to antigenic or mitogenic stimulation, this protein is required for T-cell proliferation and other activities crucial to regulation of the immune response. Can stimulate B-cells, monocytes, lymphokine-activated killer cells, natural killer cells, and glioma cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR