Anti-Staphyloccocus aureus Rosenbach tropina Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-4582R-HRP
10403-442EA 757.96 USD
10403-442
Anti-Staphyloccocus aureus Rosenbach tropina Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Staphyloccocus aureus Rosenbach tropina

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: Staphyloccocus aureus Rosenbach tropina
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR