Anti-Staphyloccocus aureus Rosenbach tropina Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-4582R-A647
10403-436EA 746.42 USD
10403-436
Anti-Staphyloccocus aureus Rosenbach tropina Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Staphyloccocus aureus Rosenbach tropina

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Staphyloccocus aureus Rosenbach tropina
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR