Anti-Listeria tropina Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-4581R-A555
10400-152EA 746.42 USD
10400-152
Anti-Listeria tropina Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Listeria tropina

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Listeria tropina
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR