Anti-VEGFA Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-4572R-CY5.5
10400-136EA 811.1 USD
10400-136
Anti-VEGFA Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Growth factor active in angiogenesis, vasculogenesis and endothelial cell growth. Induces endothelial cell proliferation, promotes cell migration, inhibits apoptosis and induces permeabilization of blood vessels. Binds to the FLT1/VEGFR1 and KDR/VEGFR2 receptors, heparan sulfate and heparin. NRP1/Neuropilin-1 binds isoforms VEGF-165 and VEGF-145. Isoform VEGF165B binds to KDR but does not activate downstream signaling pathways, does not activate angiogenesis and inhibits tumor growth.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: VEGFA
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR