Anti-ADD1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-4559R-A555
10403-276EA 811.1 USD
10403-276
Anti-ADD1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Adducins are a family of cytoskeleton proteins encoded by three genes (alpha, beta and gamma). Adducin is a protein associated with the inner leaflet of the plasma membrane and is one of the proteins localized at the spectrin-Actin junction of the membrane skeleton. The cortical Actin cytoskeletal network is lost during apoptosis and Adducins are central in the cortical Actin network organization. Adducin alpha is a cytoskeletal protein involved with sodium-pump activity in the renal tubule and is associated with hypertension. The expression of Adducin alpha and Adducin gamma is ubiquitous in contrast to the restricted expression of Adducin beta . Adducin beta is expressed at high levels in brain and hematopoietic tissues, such as bone marrow in humans and spleen in mice.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADD1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR