Anti-PE Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-4550R-CY5.5
10403-230EA 811.1 USD
10403-230
Anti-PE Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Pectinesterase

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PE
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR