Anti-PE Rabbit Polyclonal Antibody (Biotin)

BS-4550R-BIO
10403-224EA 529.29 USD
10403-224
Anti-PE Rabbit Polyclonal Antibody (Biotin)
Pectinesterase

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: PE
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR